
1H,1H,8H,8H-Dodecafluoro-1,8-octandiol ≥98.0% (by GC)
TCIAD2891-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:90.0 to 94.0 deg-C
- Solubility in Methanol:almost transparency
:
:362.12 g/mol
:GHS07
:90177-96-1
:C₈H₆F₁₂O₂
:MFCD00153145