Order Entry
Italy
ContactUsLinkComponent
5-Chloro-2-pentanone ≥98.0% (by GC) stabilised
Flammable
Irritant
5-Chloro-2-pentanone ≥98.0% (by GC) stabilised
Catalog # TCIAC0867-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
5-Chloro-2-pentanone ≥98.0% (by GC) stabilised
Catalog # TCIAC0867-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Brown clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :120.58 g/mol
Boiling Pt:106°C
Melting Pt:–48 °C
Density:1,057 g/cm³ (20 °C)
Flash Pt:35°C
Storage Temperature:Refrigerator
EINECS:227-565-8
UN:1224
ADR:3,III
 :Stabilised with potassium carbonate
 :GHS02
 :GHS07
CAS Number:5891-21-4
Formula:C₅H₉ClO
MDL Number:MFCD00001008