Order Entry
Italy
ContactUsLinkComponent
Cellulase (from Aspergillus niger)
Cellulase (from Aspergillus niger)
Supplier:  TCI
Certificates
Cellulase (from Aspergillus niger)
Supplier:  TCI
Certificates

Specifications

  • CAS No.:
    9012-54-8
  • Source:
    Aspergillus niger
  • MDL No.:
    MFCD00081510
  • Enzyme name:
    Cellulase

Specifications

Related Information

Frequently Bought Together