Order Entry
Italy
ContactUsLinkComponent
Disodium naphthalene-1,5-disulphonate ≥98.0% (by titrimetric analysis)
Disodium naphthalene-1,5-disulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0012-25G
Supplier:  TCI
CAS Number:  
undefined
Disodium naphthalene-1,5-disulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0012-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal

Specification Test Results

About this item

 :
 :332.27 g/mol
EINECS:216-732-0
CAS Number:1655-29-4
Formula:C₁₀H₆Na₂O₆S₂
MDL Number:MFCD00064178