Magnesium sulphate 1 M in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7487-88-9
:
Formula:MgSO₄
MDL Number:MFCD00011110
:120.37 g/mol
Storage Temperature:Ambient
EINECS:231-298-2
Frequently Bought Together