L-Methionine cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63-68-3
:
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063097
:149.21 g/mol
Merck Index:14,5975
Boiling Pt:278 °C (1013 hPa)
Melting Pt:284 °C (dec.)
Density:1.373 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-562-9
Related Information
Frequently Bought Together