Order Entry
Italy
ContactUsLinkComponent
 
New fuchsine for biochemical research
Catalog # TCIAN0873-5G
Supplier:  TCI
CAS Number:  
undefined
New fuchsine for biochemical research
Catalog # TCIAN0873-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Green to Dark green powder to crystal
  • Lambda max.:
    550.0 to 558.0 nm (EtOH)
  • Absorbance (E1%1cm):
    min. 1,500 (EtOH, 550.0 to 558.0 nm)
  • Alkaline Phosphatase detection test:
    to pass test

Specification Test Results

About this item

 :
 :365.91 g/mol
EINECS:221-831-7
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:3248-91-7
Formula:C₂₂H₂₄ClN₃
MDL Number:MFCD00012567