Calcium carbonate 98.0-100.5%, powder USP, FCC, J.T.Baker®
: Avantor
:471-34-1
:
:CaCO₃
:MFCD00010906
:100.09 g/mol
:13,01658
:825 °C
:2.71 g/cm³ (20 °C)
:Ambient
:207-439-9
- Bulk Pharmaceutical Chemical:98.0 - 100.5 %
- USP - Assay (CaCO₃) (dried basis):98.0 - 100.5 %
- USP - Identification:Passes Test
- USP - Acid-Insoluble Substances:≤0.2 %
- USP - Fluoride (F):≤0.005 %
- USP - Arsenic (As):≤3 ppm
- USP - Barium (Ba):Passes Test
- USP - Iron (Fe):≤0.1 %
- USP - Lead (Pb) (USP):≤3 ppm
- USP - Magnesium and Alkali Salts (as SO₄):≤1.0 %
- USP - Mercury (Hg):≤0.5 ppm
- FCC - Assay (CaCO₃) (dried basis):98.0 - 100.5 %
- FCC - Acid-Insoluble Substances:≤0.2 %
- FCC - Arsenic (As):≤3 ppm
- FCC - Fluoride (F):≤0.005 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb) (FCC):≤3 mg/kg
- FCC - Loss on Drying:≤2.0 %
- FCC - Magnesium and Alkali Salts (as SO₄):≤1.0 %
- Bulk Density (lb/cu ft):≥28
Frequently Bought Together