
4'-Hydroxyheptanophenone ≥98.0% (by GC, titration analysis)
: TCI
:14392-72-4
:
:C₁₃H₁₈O₂
:MFCD00191523
:206.2800 g/mol
:220°C
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:92.0 to 95.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together