
3-Bromobenzylamine hydrochloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:39959-54-1
:
Formula:C₇H₉BrClN
MDL Number:MFCD00012856
:222.52 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):97.0 and le 103.0 %
- Melting point:198.0 to 222.0 deg-C
Specification Test Results
Frequently Bought Together