Order Entry
Italy
ContactUsLinkComponent
(R)-(-)-2-Aminononane ≥99%, ee 98%, ChiPros®
(R)-(-)-2-Aminononane ≥99%, ee 98%, ChiPros®
  L19310.14
 :  
(R)-(-)-2-Aminononane ≥99%, ee 98%, ChiPros®
  L19310.14
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
 :74069-74-2
 :
 :C₉H₂₁N
 :MFCD03844739
 :143.27 g/mol
 :73 °C (19 mmHg)
 :0.782
 :71 °C (159 °F)
 :Ambient
 :2735
 :8,II
 :GHS05