Order Entry
Italy
ContactUsLinkComponent
Terbium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Oxidizing
Irritant
Terbium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Catalog # 40573.NC
CAS Number:  
Oxidizing
Irritant
Terbium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Catalog # 40573.NC
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

CAS Number:14014-09-6
 :
Formula:Cl₃O₁₂Tb
MDL Number:MFCD00150362
 :457.277 g/mol
Density:1.5
Storage Temperature:Ambient
EINECS:237-826-8
UN:3211
ADR:5.1,II
 :GHS03
 :GHS07