

Nickel copper alloy (67:33 w%), alloy 400, foil, Thickness: 1.6 mm (0.062 in)
Catalog # 45231.GT
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1772
- Pk Info:150×150 mm
Specifications
About this item
:
:GHS08
:GHS07
Storage Temperature:Ambient