
4-(Methylthio)phenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1073-72-9
:
Formula:C₇H₈OS
MDL Number:MFCD00002351
:140.21 g/mol
Boiling Pt:154°C
EINECS:214-031-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:82.0 to 87.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Related Information
Frequently Bought Together