
4-Butylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:145240-28-4
:
Formula:C₁₀H₁₅BO₂
MDL Number:MFCD02093926
:178.04 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
Related Information
Frequently Bought Together