2-Chlorothioxanthone
Glass bottle for solids, 25 mg
Catalog # PROOBP559
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
- CAS Number:86-39-5
- Formula:C₁₃H₇ClOS
- MDL Number:MFCD00005067
- Molecular Weight:246.72 g/mol
- EINECS:201-667-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg



