Order Entry
Italy
ContactUsLinkComponent
3-Nitrobenzylamine hydrochloride 97%
Irritant
3-Nitrobenzylamine hydrochloride 97%
Certificates
undefined
Irritant
3-Nitrobenzylamine hydrochloride 97%

Specification Test Results

  • Appearance:
    Off-white to tan Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    226 to 231 °C
  • Titration with HClO4:
    ≥96.0 %
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together