
4-(Methylsulphonyl)phenol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14763-60-1
:
Formula:C₇H₈O₃S
MDL Number:MFCD00047806
:172.20 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:93.0 to 97.0 deg-C
Specification Test Results
Frequently Bought Together