
4-(Methylsulphonyl)phenol 95%
ACRO424880025
:
- Pack type:Glass bottle
- Pk:2,5 g
- Seq:3122
- Appearance:Tan to pink-brown Powder
- Infrared spectrum:Conforms
- Melting point:90 to 95 °C
- Water:≤5.0 %
- HPLC:≥94 %
:
:172.20 g/mol
:90…95 °C
:GHS07
:Ambient
:14763-60-1
:C₇H₈O₃S
:MFCD00047806