1,1'-Azobis(N,N-dimethylformamide) ≥97.0% (by titrimetric analysis)
: TCI
:10465-78-8
:
:C₆H₁₂N₄O₂
:MFCD00008318
:172.19 g/mol
:Freezer
:233-951-7
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (Iodometric Titration):min. 97.0 %
- Melting point:110.0 to 114.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together