Cholesterol oleyl carbonate ≥70% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:17110-51-9
:
Formula:C₄₆H₈₀O₃
MDL Number:MFCD00003634
:681.125 g/mol
EINECS:241-179-7
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC) (CAD):min. 70.0 area%
- Specific rotation [a]20/D:-21.0 to -25.0 deg (C=2, CHCl3)
Specification Test Results
Frequently Bought Together