Order Entry
Italy
ContactUsLinkComponent
 
 
1,4-Thioxane 1,1-dioxide ≥98.0% (by GC)
 :  TCI
 
undefined
 
1,4-Thioxane 1,1-dioxide ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    131.0 to 135.0 deg-C

 

Frequently Bought Together