
4-(Trifluoromethylthio)benzyl bromide ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:21101-63-3
:
Formula:C₈H₆BrF₃S
MDL Number:MFCD00129189
:271.10 g/mol
Boiling Pt:118°C
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:53.0 to 57.0 deg-C
Specification Test Results
Frequently Bought Together