Capillary columns, PERMABOND® SE-54 HKW
Supplier: MACHEREY-NAGEL
Certificates
About this item
Special GC phase for the analysis of volatile halogenated hydrocarbons.
- Optimised nonpolar phase for volatile halogenated hydrocarbons
- Max. temperature for isothermal operation 300 °C
- Max. temperature for short isotherms in a temperature program 320 °C
USP code: G36.
Frequently Bought Together