SPE columns, PAH analysis, CHROMABOND® NH₂/C₁₈
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® NH2/C18 is a combination phase for SPE analysis of PAHs from water containing humic acids.
- Aminopropyl phase for removal of interfering humic acids
- Octadecyl phase for enrichment of PAHs
Application: Extraction of PAHs from water containing humic acids.
Specifications
- Catalog No:
- 516-0689
- 516-0688
- pH range:
- 2 - 8
- 2 - 8
Specifications
Frequently Bought Together