Order Entry
Italy
ContactUsLinkComponent
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) ≥98.0% (by titrimetric analysis) for biochemical research
Irritant
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) ≥98.0% (by titrimetric analysis) for biochemical research
Catalog # TCIAA2166-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) ≥98.0% (by titrimetric analysis) for biochemical research
Catalog # TCIAA2166-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Dark green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Peroxidase detection test:
    to pass test

Specification Test Results

About this item

 :
 :548.68 g/mol
EINECS:250-396-6
 :GHS07
Storage Temperature:Refrigerator
CAS Number:30931-67-0
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404