
MOPSO (3-(4-morpholino)-2-hydroxypropanesulphonic acid) ≥98.0% (by titrimetric analysis)
TCIAH0671-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Absorbance:max. 0.15 (1mol/L、H2O、260nm)
- Solubility in Water:almost transparency
- Heavy metals (as Pb):max. 5 ppm
:
:225.27 g/mol
:269-989-6
:GHS07
:Ambient
:68399-77-9
:C₇H₁₅NO₅S
:MFCD00009629