

Di-n-Alkyldimethylammonium chloride (mixture) ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:61789-80-8
:
MDL Number:MFCD01775729
EINECS:263-090-2
UN:3077
ADR:9,III
:GHS05
:GHS09
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (HPLC) (CAD):min. 93.0 % (C16 and C18 mixture)
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together