Order Entry
Italy
ContactUsLinkComponent
Glutaric acid ∼50% ∼4.3 mol/L in water
Irritant
Glutaric acid ∼50% ∼4.3 mol/L in water
Catalog # TCIAG0245-500G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Glutaric acid ∼50% ∼4.3 mol/L in water
Catalog # TCIAG0245-500G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    50.0% to 55.0 w/w%

Specification Test Results

About this item

 :
 :132.12 g/mol
Merck Index:14,04473
EINECS:203-817-2
 :GHS07
CAS Number:110-94-1
Formula:C₅H₈O₄
MDL Number:MFCD00004410