Order Entry
Italy
ContactUsLinkComponent
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16) ≥98.0% (by total nitrogen basis) for biochemical research
Irritant
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16) ≥98.0% (by total nitrogen basis) for biochemical research
Supplier:  TCI
Certificates
undefined
Irritant
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-16, SB-16) ≥98.0% (by total nitrogen basis) for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Suitability for protein-analysis:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together