




tert-Butyl hydroperoxide 70% in water
Catalog # TCIAB3153-100G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Iodometric titration):65.0 to 71.0 w/w%
Specification Test Results
About this item
:
:90.12 g/mol
Merck Index:14,01570
Boiling Pt:89°C
Melting Pt:-3°C
Melting Pt:–3°C
Flash Pt:43°C
Storage Temperature:Refrigerator
EINECS:200-915-7
UN:3109
ADR:5.2,
:GHS02
:GHS05
:GHS06
:GHS08
:GHS09
CAS Number:75-91-2
Formula:C₄H₁₀O₂
MDL Number:MFCD00002130