Dextran, M.W. ~ 40000 (Dextran 40)
Supplier: TCI
Certificates
About this item
CAS Number:9004-54-0
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935
Merck Index:14,02948
EINECS:232-677-5
Specification Test Results
- Appearance:White powder to crystal
- Specific rotation [a]20/D:+193 to +201 deg (C=6, H2O) (calcd.on dried substance)
- Solubility in hot Water:almost transparency
- Drying loss:max. 5.0 %
- limiting viscosity:0.16 to 0.19 cp (after drying)
Specification Test Results
Related Information
Frequently Bought Together