Order Entry
Italy
ContactUsLinkComponent
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥99.0% (by GC)
Irritant
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥99.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystaline
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    162.0 to 165.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together