


Sulphuryl chloride 1 M in dichloromethane
Catalog # H31899.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
CAS Number:7791-25-5
:
Formula:Cl₂O₂S
MDL Number:MFCD00011451
:134.97 g/mol
Density:1.352
Storage Temperature:Ambient
EINECS:232-245-6
UN:2922
ADR:8,III
:GHS05
:GHS08
:GHS07