Order Entry
Italy
ContactUsLinkComponent
Aluminium chloride ≥98.0% (by titrimetric analysis)
Corrosive
Aluminium chloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
Aluminium chloride ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together