
2,2'-Oxydiethanethiol ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2150-02-9
:
Formula:C₄H₁₀OS₂
MDL Number:MFCD00004889
:138.25 g/mol
Boiling Pt:217°C
Flash Pt:90°C
EINECS:218-421-5
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Purity (Iodometric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together