

DFDNB (1,5-Difluoro-2,4-dinitrobenzene) ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:327-92-4
:
Formula:C₆H₂F₂N₂O₄
MDL Number:MFCD00007052
:204.09 g/mol
Boiling Pt:132 °C (2 -rr)
Melting Pt:74…76 °C
Density:1,86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-324-0
UN:2811
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:73.0 to 76.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Related Information
Frequently Bought Together