
Potassium (4-methylphenyl)trifluoroborate ≥98.0% (by HPLC)
Catalog # TCIAP1583-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 97.0 %
Specification Test Results
About this item
:
:198.04 g/mol
Melting Pt:294°C
:GHS07
CAS Number:216434-82-1
Formula:C₇H₇BF₃K
MDL Number:MFCD01318171