Order Entry
Italy
ContactUsLinkComponent
Isoamyl acetate ≥98.0% (by GC) for spectrophotometry
Isoamyl acetate ≥98.0% (by GC) for spectrophotometry
Catalog # TCIAA0264-100ML
Supplier:  TCI
CAS Number:  
undefined
Isoamyl acetate ≥98.0% (by GC) for spectrophotometry
Catalog # TCIAA0264-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Absorbance (260nm):
    max. 0.500
  • Absorbance (270nm):
    max. 0.100
  • Absorbance (280nm):
    max. 0.050

Specification Test Results

About this item

Synonyms: Isopentyl acetate, Acetic acid isoamyl ester
 :
 :130.19 g/mol
Merck Index:14,05111
Boiling Pt:142°C
Melting Pt:–78 °C
Density:0,87 g/cm³ (20 °C)
Flash Pt:38°C
EINECS:204-662-3
UN:1104
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:123-92-2
Formula:C₇H₁₄O₂
MDL Number:MFCD00008946