Order Entry
Italy
ContactUsLinkComponent
 
 
 
2-Ethylhexane-1,3-diol (mixture of isomers)
 :  TCI
 
undefined
 
 
2-Ethylhexane-1,3-diol (mixture of isomers)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 % (total of diastereoisomers)

 

Frequently Bought Together