Order Entry
Italy
ContactUsLinkComponent
 
Acrylic acid ≥99.0% (by GC) stabilised
 :  TCI
 
undefined
Acrylic acid ≥99.0% (by GC) stabilised
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White or Colorless powder to lump to clear liquid
  • Purity (GC):
    min. 99.0 %

 

Frequently Bought Together