
(1S)-(+)-Camphor-10-sulphonic acid ≥98.0% (by titrimetric analysis)
: TCI
:3144-16-9
:
:C₁₀H₁₆O₄S
:MFCD00064157
:232.3 g/mol
:14,01734
:375 °C (1013 hPa)
:200°C
:221-554-1
:3261
:8,III
:GHS05
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+20.0 to +23.0 deg (C=5, H2O)
- Solubility in Water:almost transparency
Frequently Bought Together