



Ethylamine ∼70% ∼12 mol/L in water
Supplier: TCI
Certificates
About this item
CAS Number:75-04-7
:
Formula:C₂H₇N
MDL Number:MFCD00008160
:45.08 g/mol
Merck Index:14,03762
Flash Pt:–24°C
Storage Temperature:Refrigerator
EINECS:200-834-7
UN:2270
ADR:3,II
:GHS02
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):69.0 to 73.0 w/w%
Specification Test Results
Frequently Bought Together