Order Entry
Italy
ContactUsLinkComponent
Ethyl N-acetyl-N-butyl-β-alaninate
Ethyl N-acetyl-N-butyl-β-alaninate
Catalog # EHERC10929010
Supplier:  EHRENSTORFER
CAS Number:  
Ethyl N-acetyl-N-butyl-β-alaninate
Catalog # EHERC10929010
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :215.29 g/mol
Boiling Pt:292 °C (1013 hPa)
Density:0.998 g/cm³ (20 °C)
Flash Pt:159 °C
EINECS:257-835-0
CAS Number:52304-36-6
Formula:C₁₁H₂₁NO₃
MDL Number:MFCD00214076
Boiling Pt:292 °C (1013 hPa)
Flash Pt:159 °C