
3-Aminophenylboronic acid monohydrate (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:206658-89-1
:
Formula:C₆H₈BNO₂·1H₂O
MDL Number:MFCD00149554
:154.96 g/mol
EINECS:250-189-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):97.0 to 116.0 %
Specification Test Results
Related Information
Frequently Bought Together