
Sodium deoxycholate ≥98.0% (by titrimetric analysis) for electrophoresis
Catalog # TCIAD1820-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+42.0 to +45.0 deg (C=2, H2O)
- Functionality test:to pass test
Specification Test Results
About this item
:
:414.56 g/mol
Merck Index:14,02899
Melting Pt:357…365 °C
EINECS:206-132-7
:GHS07
Storage Temperature:Ambient
CAS Number:302-95-4
Formula:C₂₄H₃₉NaO₄
MDL Number:MFCD00064139