


α-Bromo-p-toluonitrile ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17201-43-3
:
Formula:C₈H₆BrN
MDL Number:MFCD00001829
:196.05 g/mol
Boiling Pt:143°C
Melting Pt:115 °C
EINECS:241-246-0
UN:3261
ADR:8,II
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:115.0 to 118.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together