
(Trifluoromethyl)trimethylsilane ≥97.0% (by GC) trifluoromethylating reagent
Supplier: TCI
Certificates
About this item
CAS Number:81290-20-2
:
Formula:C₄H₉F₃Si
MDL Number:MFCD00145454
:142.2 g/mol
Merck Index:14,09683
Boiling Pt:55°C
Density:0,95 g/cm³ (20 °C)
Flash Pt:–32°C
UN:1993
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together