

Phenazone ≥99.0% (by GC, titration analysis)
: TCI
:60-80-0
:
:C₁₁H₁₂N₂O
:MFCD00003146
:188.23 g/mol
:14,00716
:319°C
:152…154 °C
:1,25 g/cm³ (20 °C)
:Ambient
:200-486-6
:GHS07
:GHS08
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:109.0 to 113.0 deg-C
- Solubility in Water:almost transparency
Frequently Bought Together