

1-Cetylpyridinium chloride monohydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6004-24-6
:
Formula:C₂₁H₃₈ClN·1H₂O
MDL Number:MFCD00149977
:358.01 g/mol
Merck Index:14,02032
Melting Pt:77…83 °C
Storage Temperature:Ambient
EINECS:204-593-9
UN:2811
ADR:6.1,II
:GHS06
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Melting point:81.0 to 86.0 deg-C
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together